Assay Details
Target Gene Details
Entrez Gene ID: | 134510 |
Gene Name: | ubiquitin like domain containing CTD phosphatase 1 |
Gene Aliases: |
CPUB1 |
Location: |
Chr.5:159263081-159286040 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBLCP1 | NM_145049.3 | NP_659486.2 | ||
AK057996.1 | BAB71628.1 | |||
AY444562.1 | AAS68538.1 | |||
BC013425.2 | AAH13425.2 | |||
DA930581.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |