Assay Details
Target Gene Details
Entrez Gene ID: | 7903 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST1, SIAT8D, ST8SIA-IV |
Location: |
Chr.5:100806935-100903285 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ST8SIA4 | NM_005668.5 | NP_005659.1 | ||
XM_011543630.1 | XP_011541932.1 | |||
AK292872.1 | ||||
BC053657.1 | AAH53657.1 | |||
L41680.1 | AAC41775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3890678 | Chr.5:100324445 - 101099725 on Build GRCh38 | Gain | ST8SIA4 MIR548P FAM174A LOC100133050 |
nsv1023765 | Chr.5:100579307 - 101398051 on Build GRCh38 | Loss | ST8SIA4 MIR548P FAM174A |
nsv1021183 | Chr.5:100290153 - 100885722 on Build GRCh38 | Gain | ST8SIA4 MIR548P FAM174A LOC100133050 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |