Assay Details
Target Gene Details
Entrez Gene ID: | 10699 |
Gene Name: | corin, serine peptidase |
Gene Aliases: |
ATC2, CRN, Lrp4, PEE5, TMPRSS10 |
Location: |
Chr.4:47593998-47838106 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORIN | NM_001278585.1 | NP_001265514.1 | ||
NM_001278586.1 | NP_001265515.1 | |||
NM_006587.3 | NP_006578.2 | |||
AF133845.1 | AAD31850.1 | |||
AK297675.1 | ||||
AK304038.1 | ||||
AK304466.1 | ||||
BC110451.2 | AAI10452.1 | |||
BX648285.1 |
Target Gene Details
Entrez Gene ID: | 101927179 |
Gene Name: | uncharacterized LOC101927179 |
Gene Aliases: |
- |
Location: |
Chr.4:47831345-47896848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927179 | NR_125880.1 | |||
NR_125881.1 | ||||
AL599247.1 | ||||
BX280044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5202n100 | Chr.4:47486414 - 47904818 on Build GRCh38 | Gain | ATP10D MIR8053 NFXL1 LOC101927179 CORIN |
nsv4328 | Chr.4:47814450 - 47860236 on Build GRCh38 | Deletion | NFXL1 LOC101927179 CORIN |
nsv594118 | Chr.4:47558369 - 47915713 on Build GRCh38 | Loss | LOC101927157 ATP10D MIR8053 NFXL1 LOC101927179 CORIN |
nsv829924 | Chr.4:47668888 - 47864648 on Build GRCh38 | Gain | NFXL1 LOC101927179 CORIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |