Assay Details
Target Gene Details
Entrez Gene ID: | 53343 |
Gene Name: | nudix hydrolase 9 |
Gene Aliases: |
NUDT10 |
Location: |
Chr.4:87420002-87459454 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT9 | NM_001248011.1 | NP_001234940.1 | ||
NM_024047.4 | NP_076952.1 | |||
NM_198038.2 | NP_932155.1 | |||
XM_011532032.2 | XP_011530334.1 | |||
AF265341.1 | AAM46066.1 | |||
AF273028.1 | AAM46068.1 | |||
AK027295.1 | BAB55021.1 | |||
AK074845.1 | BAC11239.1 | |||
AK075408.1 | BAC11601.1 | |||
AK308028.1 | ||||
AY026252.1 | AAK07671.1 | |||
AY359123.1 | AAQ89480.1 | |||
BC000542.2 | AAH00542.1 | |||
BM544951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3601276 | Chr.4:87422870 - 87515781 on Build GRCh38 | Loss | SPARCL1 NUDT9 |
nsv1011882 | Chr.4:87442807 - 87481325 on Build GRCh38 | Gain | SPARCL1 NUDT9 |
nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss | HSD17B11 HSD17B13 SPARCL1 NUDT9 DMP1 DSPP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |