Assay Details
Target Gene Details
Entrez Gene ID: | 1499 |
Gene Name: | catenin beta 1 |
Gene Aliases: |
CTNNB, MRD19, armadillo |
Location: |
Chr.3:41199451-41240448 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTNNB1 | NM_001098209.1 | NP_001091679.1 | ||
NM_001098210.1 | NP_001091680.1 | |||
NM_001904.3 | NP_001895.1 | |||
XM_005264886.2 | XP_005264943.1 | |||
XM_006712985.1 | XP_006713048.1 | |||
XM_017005738.1 | XP_016861227.1 | |||
AK289932.1 | ||||
AK294778.1 | ||||
AK296790.1 | ||||
AK299954.1 | ||||
AK314776.1 | ||||
BC058926.1 | AAH58926.1 | |||
DA216720.1 | ||||
X87838.1 | CAA61107.1 | |||
Z19054.1 | CAA79497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954468 | Chr.3:41200310 - 41224409 on Build GRCh38 | Duplication | CTNNB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |