Assay Details
Target Gene Details
Entrez Gene ID: | 80218 |
Gene Name: | N(alpha)-acetyltransferase 50, NatE catalytic subunit |
Gene Aliases: |
MAK3, NAT13, NAT13P, NAT5, NAT5P, SAN |
Location: |
Chr.3:113716460-113746299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA50 | NM_001308445.1 | NP_001295374.1 | ||
NM_025146.3 | NP_079422.1 | |||
AK023090.1 | BAB14397.1 | |||
AK023256.1 | BAB14490.1 | |||
AK296456.1 | ||||
AK307184.1 | ||||
AL601126.1 | ||||
BC012731.2 | AAH12731.1 | |||
BM831426.1 | ||||
CR749314.1 | CAH18169.1 | |||
DA854531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain | ZDHHC23 SPICE1 CFAP44 GRAMD1C MIR4446 ATP6V1A BOC SIDT1 CFAP44-AS1 MIR8076 USF3 NAA50 |
nsv829678 | Chr.3:113727330 - 113914206 on Build GRCh38 | Loss | GRAMD1C ATP6V1A NAA50 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |