Assay Details
Target Gene Details
Entrez Gene ID: | 6599 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 |
Gene Aliases: |
BAF155, CRACC1, Rsc8, SRG3, SWI3 |
Location: |
Chr.3:47585888-47781915 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMARCC1 | NM_003074.3 | NP_003065.3 | ||
XM_011534034.1 | XP_011532336.1 | |||
AK302412.1 | ||||
AK302541.1 | ||||
BC050564.1 | AAH50564.1 | |||
BC113465.1 | ||||
BC117213.1 | ||||
U66615.1 | AAC50693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3807 | Chr.3:47558395 - 47607814 on Build GRCh38 | Insertion | CSPG5 SMARCC1 |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss | PRSS42 SETD2 CCDC12 TMIE KLHL18 KIF9 PTH1R SNORD13P3 PRSS50 PTPN23 CSPG5 PRSS45 PRSS46 SCAP KIF9-AS1 ALS2CL NRADDP ELP6 NBEAL2 SMARCC1 MYL3 PRSS43 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |