Assay Details
Target Gene Details
Entrez Gene ID: | 90288 |
Gene Name: | EF-hand calcium binding domain 12 |
Gene Aliases: |
C3orf25 |
Location: |
Chr.3:129401321-129428651 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB12 | NM_207307.2 | NP_997190.1 | ||
XM_011513293.2 | XP_011511595.1 | |||
XM_011513294.2 | XP_011511596.1 | |||
XM_011513295.2 | XP_011511597.1 | |||
XM_011513297.2 | XP_011511599.1 | |||
XM_011513298.1 | XP_011511600.1 | |||
XM_011513299.2 | XP_011511601.1 | |||
AK096099.1 | ||||
AK127968.1 | ||||
AL133011.1 | CAH10712.1 | |||
BC066975.1 | AAH66975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2725901 | Chr.3:129380821 - 129487939 on Build GRCh38 | Deletion | RPL32P3 EFCAB12 IFT122 SNORA7B MBD4 |
esv3597741 | Chr.3:129383049 - 129486193 on Build GRCh38 | Loss | RPL32P3 EFCAB12 IFT122 SNORA7B MBD4 |
nsv591668 | Chr.3:129403449 - 129490335 on Build GRCh38 | Gain | EFCAB12 IFT122 MBD4 |
nsv591652 | Chr.3:129327063 - 129408132 on Build GRCh38 | Gain | RPL32P3 EFCAB12 SNORA7B |
esv3597742 | Chr.3:129400638 - 129408733 on Build GRCh38 | Loss | EFCAB12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |