Assay Details
Target Gene Details
Entrez Gene ID: | 54918 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
CKLFSF6, PRO2219 |
Location: |
Chr.3:32481312-32502911 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM6 | NM_017801.2 | NP_060271.1 | ||
AF479261.1 | AAN73039.1 | |||
AK000403.1 | BAA91141.1 | |||
BC002797.2 | AAH02797.1 | |||
CR457231.1 | CAG33512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain | GLB1 SUSD5 CCR4 OSBPL10-AS1 LOC105377022 LOC105377021 MIR548AY CMTM7 OSBPL10 ZNF860 DYNC1LI1 CMTM6 GPD1L CRTAP TMPPE STT3B CMTM8 TRIM71 CNOT10 |
esv3595776 | Chr.3:32495893 - 32501883 on Build GRCh38 | Loss | CMTM6 |
nsv834652 | Chr.3:32375792 - 32564922 on Build GRCh38 | Loss | DYNC1LI1 CMTM6 MIR548AY CMTM7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |