Assay Details
Target Gene Details
Entrez Gene ID: | 9117 |
Gene Name: | SEC22 homolog C, vesicle trafficking protein |
Gene Aliases: |
SEC22L3 |
Location: |
Chr.3:42547967-42601080 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEC22C | NM_001201572.1 | NP_001188501.1 | ||
NM_001201584.1 | NP_001188513.1 | |||
NM_004206.3 | NP_004197.1 | |||
NM_032970.3 | NP_116752.1 | |||
XM_005265533.1 | XP_005265590.1 | |||
XM_005265534.1 | XP_005265591.1 | |||
XM_005265535.1 | XP_005265592.1 | |||
XM_011534212.1 | XP_011532514.1 | |||
XM_017007438.1 | XP_016862927.1 | |||
XM_017007439.1 | XP_016862928.1 | |||
AK074576.1 | ||||
AK292856.1 | ||||
AK293133.1 | ||||
AY359041.1 | AAQ89400.1 | |||
BC006178.1 | AAH06178.1 | |||
BC018437.1 | AAH18437.1 | |||
BX377531.2 | ||||
DA434403.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)