Assay Details
Target Gene Details
Entrez Gene ID: | 54585 |
Gene Name: | leucine zipper transcription factor like 1 |
Gene Aliases: |
BBS17 |
Location: |
Chr.3:45823316-45916042 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LZTFL1 | NM_001276378.1 | NP_001263307.1 | ||
NM_001276379.1 | NP_001263308.1 | |||
NM_020347.3 | NP_065080.1 | |||
NR_075080.1 | ||||
XM_006713207.2 | XP_006713270.2 | |||
XM_011533838.2 | XP_011532140.1 | |||
XM_017006644.1 | XP_016862133.1 | |||
XM_017006645.1 | XP_016862134.1 | |||
AJ297351.1 | CAB95836.1 | |||
AK024323.1 | ||||
AK093705.1 | ||||
AK303416.1 | ||||
BC025988.2 | AAH25988.1 | |||
BC042483.1 | AAH42483.1 | |||
BX640604.1 | CAE45710.1 | |||
CN429018.1 | ||||
DA408809.1 | ||||
DC410047.1 | ||||
HY159348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain | CXCR6 LZTFL1 FYCO1 SLC6A20 CCR9 |
nsv3796 | Chr.3:45809469 - 45844553 on Build GRCh38 | Deletion | LZTFL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |