Assay Details
Target Gene Details
Entrez Gene ID: | 545 |
Gene Name: | ATR serine/threonine kinase |
Gene Aliases: |
FCTCS, FRP1, MEC1, SCKL, SCKL1 |
Location: |
Chr.3:142449235-142578826 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATR | NM_001184.3 | NP_001175.2 | ||
XM_011512924.1 | XP_011511226.1 | |||
XM_011512925.1 | XP_011511227.1 | |||
XM_011512926.2 | XP_011511228.1 | |||
XM_017006643.1 | XP_016862132.1 | |||
U49844.1 | AAC50405.1 | |||
U76308.1 | AAC50929.1 | |||
Y09077.1 | CAA70298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523945 | Chr.3:142214807 - 142500700 on Build GRCh38 | Gain | XRN1 GK5 ATR |
nsv528954 | Chr.3:142445175 - 142500700 on Build GRCh38 | Loss | XRN1 ATR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |