Assay Details
Target Gene Details
Entrez Gene ID: | 10015 |
Gene Name: | programmed cell death 6 interacting protein |
Gene Aliases: |
AIP1, ALIX, DRIP4, HP95 |
Location: |
Chr.3:33798571-33869707 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDCD6IP | NM_001162429.2 | NP_001155901.1 | ||
NM_001256192.1 | NP_001243121.1 | |||
NM_013374.5 | NP_037506.2 | |||
XM_011533252.1 | XP_011531554.1 | |||
AF151793.1 | AAF08220.1 | |||
AF250394.1 | AAQ14260.1 | |||
AF349951.1 | AAK20398.1 | |||
BC020066.1 | AAH20066.1 | |||
BC068454.1 | AAH68454.1 | |||
BT007367.1 | AAP36031.1 | |||
DA652213.1 | ||||
GQ131806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522881 | Chr.3:33694634 - 34200194 on Build GRCh38 | Gain | CLASP2 PDCD6IP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |