Assay Details
Target Gene Details
Entrez Gene ID: | 25937 |
Gene Name: | WW domain containing transcription regulator 1 |
Gene Aliases: |
TAZ |
Location: |
Chr.3:149517235-149724783 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WWTR1 | NM_001168278.1 | NP_001161750.1 | ||
NM_001168280.1 | NP_001161752.1 | |||
NM_015472.4 | NP_056287.1 | |||
XM_017006121.1 | XP_016861610.1 | |||
XM_017006122.1 | XP_016861611.1 | |||
AJ299431.1 | CAC17722.1 | |||
AK022036.1 | BAB13957.1 | |||
AL050107.1 | CAB43275.1 | |||
AL833852.1 | CAD38711.1 | |||
BC014052.2 | AAH14052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893785 | Chr.3:149507931 - 149583818 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)