Assay Details
Target Gene Details
Entrez Gene ID: | 7709 |
Gene Name: | zinc finger and BTB domain containing 17 |
Gene Aliases: |
MIZ-1, ZNF151, ZNF60, pHZ-67 |
Location: |
Chr.1:15941869-15976132 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB17 | NM_001242884.1 | NP_001229813.1 | ||
NM_001287603.1 | NP_001274532.1 | |||
NM_001287604.1 | NP_001274533.1 | |||
NM_001324137.1 | NP_001311066.1 | |||
NM_001324138.1 | NP_001311067.1 | |||
NM_003443.2 | NP_003434.2 | |||
XM_005245986.1 | XP_005246043.1 | |||
XM_005245987.1 | XP_005246044.1 | |||
XM_011542085.2 | XP_011540387.1 | |||
XM_011542086.1 | XP_011540388.1 | |||
XM_011542087.2 | XP_011540389.1 | |||
XM_011542088.1 | XP_011540390.1 | |||
AK223618.1 | BAD97338.1 | |||
AK226168.1 | ||||
AK294796.1 | ||||
AK299820.1 | ||||
AK301896.1 | ||||
AK302610.1 | ||||
AK303823.1 | ||||
AK309081.1 | ||||
AK311083.1 | ||||
AK315201.1 | ||||
BC126163.1 | ||||
BC143965.1 | ||||
DB064089.1 | ||||
Y09723.1 | CAA70889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508936 | Chr.1:15915852 - 16181025 on Build GRCh38 | Insertion | SPEN FAM131C C1orf64 HSPB7 EPHA2 CLCNKA ZBTB17 CLCNKB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |