Assay Details
Target Gene Details
Entrez Gene ID: | 22920 |
Gene Name: | kinesin associated protein 3 |
Gene Aliases: |
FLA3, KAP-1, KAP-3, KAP3, SMAP, Smg-GDS, dJ190I16.1 |
Location: |
Chr.1:169921329-170074741 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIFAP3 | NM_001204514.1 | NP_001191443.1 | ||
NM_001204516.1 | NP_001191445.1 | |||
NM_001204517.1 | NP_001191446.1 | |||
NM_014970.3 | NP_055785.2 | |||
XM_005244970.1 | XP_005245027.1 | |||
XM_011509307.2 | XP_011507609.2 | |||
AK026471.1 | ||||
AK122897.1 | ||||
AK297368.1 | ||||
AK301912.1 | ||||
AK303052.1 | ||||
AK315586.1 | ||||
BC028679.2 | AAH28679.1 | |||
U59919.1 | AAC50788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv460n100 | Chr.1:169921401 - 169960441 on Build GRCh38 | Loss |
![]() |
dgv462n100 | Chr.1:169923520 - 170000303 on Build GRCh38 | Loss |
![]() |
dgv461n100 | Chr.1:169923143 - 169981200 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)