Assay Details
Target Gene Details
Entrez Gene ID: | 100996570 |
Gene Name: | NFIA antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.1:60940244-60970776 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIA-AS2 | NR_110617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012657 | Chr.1:60299584 - 61316101 on Build GRCh38 | Gain | NFIA-AS2 NFIA LOC105378763 LOC101926964 NFIA-AS1 |