Assay Details
Target Gene Details
Entrez Gene ID: | 6491 |
Gene Name: | SCL/TAL1 interrupting locus |
Gene Aliases: |
MCPH7, SIL |
Location: |
Chr.1:47250139-47314787 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822531 | Chr.1:47248220 - 47264411 on Build GRCh38 | Gain |
![]() |
nsv1160846 | Chr.1:47249998 - 47266706 on Build GRCh38 | Duplication |
![]() |
nsv441692 | Chr.1:47247989 - 47263416 on Build GRCh38 | Gain |
![]() |
esv3578138 | Chr.1:47247976 - 47263403 on Build GRCh38 | Gain |
![]() |
esv3585954 | Chr.1:47248325 - 47258549 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)