Assay Details
Target Gene Details
Entrez Gene ID: | 113451 |
Gene Name: | antizyme inhibitor 2 |
Gene Aliases: |
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp |
Location: |
Chr.1:33081110-33162288 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576805 | Chr.1:32896710 - 33106481 on Build GRCh38 | Gain | AZIN2 AK2 RNF19B TMEM54 |
nsv834602 | Chr.1:32947691 - 33116221 on Build GRCh38 | Loss | AZIN2 AK2 RNF19B |
nsv545962 | Chr.1:32982619 - 33129611 on Build GRCh38 | Loss | AZIN2 AK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |