Assay Details
Target Gene Details
Entrez Gene ID: | 101929464 |
Gene Name: | uncharacterized LOC101929464 |
Gene Aliases: |
- |
Location: |
Chr.1:33350352-33363245 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929464 | NR_125978.1 | |||
AW072537.1 |
Target Gene Details
Entrez Gene ID: | 1912 |
Gene Name: | polyhomeotic homolog 2 |
Gene Aliases: |
EDR2, HPH2, PH2 |
Location: |
Chr.1:33323623-33431017 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHC2 | NM_198040.2 | NP_932157.1 | ||
XM_011540876.2 | XP_011539178.1 | |||
XM_011540877.2 | XP_011539179.1 | |||
XM_011540878.2 | XP_011539180.1 | |||
XM_017000513.1 | XP_016856002.1 | |||
XM_017000514.1 | XP_016856003.1 | |||
XM_017000515.1 | XP_016856004.1 | |||
XM_017000516.1 | XP_016856005.1 | |||
XM_017000517.1 | XP_016856006.1 | |||
XM_017000518.1 | XP_016856007.1 | |||
XM_017000519.1 | XP_016856008.1 | |||
XM_017000520.1 | XP_016856009.1 | |||
XM_017000521.1 | XP_016856010.1 | |||
XM_017000522.1 | XP_016856011.1 | |||
AJ419231.1 | CAD11673.1 | |||
AK292905.1 | ||||
AK294426.1 | ||||
BC130630.1 | ||||
BC144121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834613 | Chr.1:33334605 - 33496188 on Build GRCh38 | Gain | PHC2 ZSCAN20 LOC101929464 |
nsv522364 | Chr.1:33266591 - 33367332 on Build GRCh38 | Loss | PHC2 LOC101929464 MIR3605 ZNF362 A3GALT2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |