Assay Details
Target Gene Details
Entrez Gene ID: | 1939 |
Gene Name: | eukaryotic translation initiation factor 2D |
Gene Aliases: |
HCA56, LGTN |
Location: |
Chr.1:206572485-206612551 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2D |
Target Gene Details
Entrez Gene ID: | 83593 |
Gene Name: | Ras association domain family member 5 |
Gene Aliases: |
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3 |
Location: |
Chr.1:206507530-206589284 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RASSF5 | NM_182663.3 | NP_872604.1 | ||
NM_182664.3 | NP_872605.1 | |||
NM_182665.3 | NP_872606.1 | |||
AB209145.1 | BAD92382.1 | |||
AF445801.1 | AAL38592.1 | |||
AK289861.1 | ||||
AK291268.1 | ||||
AK292039.1 | ||||
AL832784.1 | CAI46164.1 | |||
AY062002.1 | AAL40388.1 | |||
AY062003.1 | AAL40389.1 | |||
AY216268.1 | AAO61668.1 | |||
AY261332.1 | AAP83360.1 | |||
BC004270.1 | AAH04270.1 | |||
BC007203.1 | AAH07203.1 | |||
BC042651.1 | AAH42651.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |