Assay Details
Target Gene Details
Entrez Gene ID: | 23341 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C16 |
Gene Aliases: |
- |
Location: |
Chr.1:15526694-15571733 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJC16 | NM_001287811.1 | NP_001274740.1 | ||
NM_015291.3 | NP_056106.1 | |||
NR_109898.1 | ||||
AB023179.2 | ||||
AK091848.1 | ||||
AK292479.1 | ||||
AL833299.1 | ||||
BC047363.1 | AAH47363.1 | |||
BX648982.1 | ||||
CR749259.1 | CAH18115.1 | |||
CR749567.1 | CAH18363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss | LOC101927441 FHAD1 CASP9 DNAJC16 CTRC EFHD2 CELA2B CELA2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |