Assay Details
Target Gene Details
Entrez Gene ID: | 56950 |
Gene Name: | SET and MYND domain containing 2 |
Gene Aliases: |
HSKM-B, KMT3C, ZMYND14 |
Location: |
Chr.1:214281101-214337134 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMYD2 | NM_020197.2 | NP_064582.2 | ||
AF226053.1 | AAF86953.1 | |||
AK313868.1 | ||||
BC028104.1 | ||||
BC098133.1 | AAH98133.1 | |||
BC098276.1 | AAH98276.1 | |||
BC098335.1 | AAH98335.1 | |||
BG723585.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |