Assay Details
Target Gene Details
Entrez Gene ID: | 60484 |
Gene Name: | hyaluronan and proteoglycan link protein 2 |
Gene Aliases: |
BRAL1 |
Location: |
Chr.1:156601504-156626608 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAPLN2 | XM_017002020.1 | XP_016857509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss | GPATCH4 BCAN CRABP2 TTC24 HAPLN2 NES NAXE LOC101928177 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |