Assay Details
Target Gene Details
Entrez Gene ID: | 2959 |
Gene Name: | general transcription factor IIB |
Gene Aliases: |
TF2B, TFIIB |
Location: |
Chr.1:88852638-88891944 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2B | NM_001514.5 | NP_001505.1 | ||
XM_011541299.1 | XP_011539601.1 | |||
AB451296.1 | ||||
AK289822.1 | ||||
BC020597.1 | AAH20597.1 | |||
BP379342.1 | ||||
F07886.1 | ||||
M76766.1 | AAA61149.1 | |||
S44184.1 | AAB23144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830537 | Chr.1:88790384 - 88940395 on Build GRCh38 | Gain | PKN2 KYAT3 GTF2B |
nsv830525 | Chr.1:88776863 - 88975647 on Build GRCh38 | Loss | PKN2 KYAT3 GTF2B |
nsv830514 | Chr.1:88715143 - 88876830 on Build GRCh38 | Loss | PKN2 GTF2B |
dgv22n27 | Chr.1:88791695 - 88894837 on Build GRCh38 | Gain | PKN2 GTF2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |