Assay Details
Target Gene Details
Entrez Gene ID: | 6705 |
Gene Name: | small proline rich protein 2F |
Gene Aliases: |
- |
Location: |
Chr.1:153112121-153119460 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPRR2F | XM_017002176.1 | XP_016857665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv547920 | Chr.1:153060930 - 153126869 on Build GRCh38 | Loss | SPRR2F SPRR2E SPRR2B |
nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain | LCE1B SPRR1B SPRR2D SPRR2E SPRR3 C1orf68 LCE6A LCE1D SPRR2F SPRR2G SPRR4 KPRP LELP1 SMCP SPRR2A LOC101928009 LINC01527 LCE1C PRR9 LCE1E LOR LCE4A LCE1F SPRR1A IVL LCE1A SPRR2C SPRR2B |
nsv1004168 | Chr.1:153115733 - 153253557 on Build GRCh38 | Loss | PRR9 SPRR2F SPRR2G LOC101928009 LELP1 SPRR2C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |