Assay Details
Target Gene Details
Entrez Gene ID: | 3155 |
Gene Name: | 3-hydroxymethyl-3-methylglutaryl-CoA lyase |
Gene Aliases: |
HL |
Location: |
Chr.1:23801877-23825459 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMGCL | NM_000191.2 | NP_000182.2 | ||
NM_001166059.1 | NP_001159531.1 | |||
AK122801.1 | ||||
AK300733.1 | ||||
AK309129.1 | ||||
AK313869.1 | ||||
BC010570.1 | AAH10570.1 | |||
BT009792.1 | AAP88794.1 | |||
CR456884.1 | CAG33165.1 | |||
FJ472654.1 | ||||
GU433941.1 | ||||
L07033.1 | AAA92733.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)