Assay Details
Target Gene Details
Entrez Gene ID: | 943 |
Gene Name: | TNF receptor superfamily member 8 |
Gene Aliases: |
CD30, D1S166E, Ki-1 |
Location: |
Chr.1:12063318-12144213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFRSF8 | NM_001243.4 | NP_001234.3 | ||
NM_001281430.2 | NP_001268359.2 | |||
XM_011542441.2 | XP_011540743.1 | |||
BC136400.1 | ||||
M83554.1 | AAA51947.1 | |||
S75768.1 | AAD14188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469830 | Chr.1:12039669 - 12199858 on Build GRCh38 | Loss | MIR7846 MIR4632 TNFRSF1B TNFRSF8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |