Assay Details
Target Gene Details
Entrez Gene ID: | 5293 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta |
Gene Aliases: |
APDS, IMD14, P110DELTA, PI3K, p110D |
Location: |
Chr.1:9629889-9729114 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3CD | XM_017001477.1 | XP_016856966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508847 | Chr.1:9612137 - 9678831 on Build GRCh38 | Insertion | PIK3CD-AS2 PIK3CD TMEM201 LOC107985734 PIK3CD-AS1 |
nsv1013224 | Chr.1:9380981 - 9639710 on Build GRCh38 | Gain | LOC100506022 PIK3CD TMEM201 LOC107985734 RPL9P11 SLC25A33 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |