Assay Details
Target Gene Details
Entrez Gene ID: | 2268 |
Gene Name: | FGR proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
SRC2, c-fgr, c-src2, p55-Fgr, p55c-fgr, p58-Fgr, p58c-fgr |
Location: |
Chr.1:27612289-27635561 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGR | NM_005248.2 | NP_005239.1 | ||
XM_006710452.2 | XP_006710515.1 | |||
XM_011541010.1 | XP_011539312.1 | |||
XM_011541011.2 | XP_011539313.1 | |||
XM_011541012.1 | XP_011539314.1 | |||
XM_011541013.2 | XP_011539315.1 | |||
XM_017000673.1 | XP_016856162.1 | |||
XM_017000674.1 | XP_016856163.1 | |||
AK130188.1 | ||||
BC064382.1 | AAH64382.1 | |||
DA139476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508700 | Chr.1:27556643 - 27653685 on Build GRCh38 | Deletion | FGR AHDC1 |
nsv522842 | Chr.1:27522789 - 27685343 on Build GRCh38 | Gain | IFI6 FGR AHDC1 |
nsv1014991 | Chr.1:27560502 - 27628874 on Build GRCh38 | Gain | FGR AHDC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |