Assay Details
Target Gene Details
Entrez Gene ID: | 22850 |
Gene Name: | ADNP homeobox 2 |
Gene Aliases: |
ZNF508 |
Location: |
Chr.18:80109031-80140346 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADNP2 | NM_014913.3 | NP_055728.1 | ||
XM_005266656.4 | XP_005266713.1 | |||
XM_011525883.2 | XP_011524185.1 | |||
AB020670.1 | ||||
AK292566.1 | ||||
BC071589.1 | AAH71589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3413n100 | Chr.18:80023442 - 80257174 on Build GRCh38 | Gain | ADNP2 RBFA TXNL4A RBFADN PARD6G PARD6G-AS1 |
nsv577958 | Chr.18:79925520 - 80130825 on Build GRCh38 | Gain | ADNP2 RBFA TXNL4A PQLC1 RBFADN HSBP1L1 |
nsv428359 | Chr.18:79277548 - 80255221 on Build GRCh38 | Gain | KCNG2 ATP9B TXNL4A RBFADN HSBP1L1 PARD6G-AS1 ADNP2 LOC105376875 NFATC1 LOC107985149 RBFA LOC284240 PQLC1 CTDP1 LOC284241 PARD6G |
nsv1065902 | Chr.18:78538045 - 80257174 on Build GRCh38 | Gain | KCNG2 ATP9B TXNL4A RBFADN HSBP1L1 SALL3 PARD6G-AS1 ADNP2 LOC105376875 NFATC1 LOC107985149 RBFA LOC284240 PQLC1 CTDP1 LOC100996274 LOC284241 PARD6G |
esv3643330 | Chr.18:80099479 - 80182196 on Build GRCh38 | Loss | ADNP2 PARD6G PARD6G-AS1 |
esv275287 | Chr.18:80118818 - 80133560 on Build GRCh38 | Gain+Loss | ADNP2 |
nsv1061450 | Chr.18:79703531 - 80135801 on Build GRCh38 | Gain | ADNP2 KCNG2 RBFA TXNL4A PQLC1 CTDP1 RBFADN HSBP1L1 |
dgv3414n100 | Chr.18:80110621 - 80257174 on Build GRCh38 | Gain | ADNP2 PARD6G PARD6G-AS1 |
esv3643326 | Chr.18:79799861 - 80157032 on Build GRCh38 | Loss | ADNP2 KCNG2 RBFA TXNL4A PQLC1 RBFADN HSBP1L1 PARD6G-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |