Assay Details
Target Gene Details
Entrez Gene ID: | 6760 |
Gene Name: | SS18, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
SSXT, SYT |
Location: |
Chr.18:26016253-26091217 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SS18 | NM_001007559.2 | NP_001007560.1 | ||
NM_001308201.1 | NP_001295130.1 | |||
NM_005637.3 | NP_005628.2 | |||
XM_006722527.2 | XP_006722590.1 | |||
XM_011526145.1 | XP_011524447.1 | |||
XM_011526147.2 | XP_011524449.1 | |||
XM_011526148.2 | XP_011524450.1 | |||
XM_011526149.2 | XP_011524451.1 | |||
XM_011526150.2 | XP_011524452.1 | |||
XM_011526151.2 | XP_011524453.1 | |||
XM_011526152.2 | XP_011524454.1 | |||
AF343880.1 | AAK21314.1 | |||
AK289510.1 | ||||
AK296949.1 | ||||
AK299082.1 | ||||
AK301504.1 | ||||
AK304305.1 | ||||
AK316302.1 | ||||
AK316338.1 | ||||
BC096221.1 | AAH96221.1 | |||
BC096222.1 | AAH96222.1 | |||
BC096223.1 | AAH96223.1 | |||
BC096224.1 | AAH96224.1 | |||
CR542088.1 | CAG46885.1 | |||
CR542103.1 | CAG46900.1 | |||
X79201.1 | CAA55792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893078 | Chr.18:25391998 - 26039471 on Build GRCh38 | Gain | SS18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |