Assay Details
Target Gene Details
Entrez Gene ID: | 26040 |
Gene Name: | SET binding protein 1 |
Gene Aliases: |
MRD29, SEB |
Location: |
Chr.18:44680173-45068510 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SETBP1 | NM_001130110.1 | NP_001123582.1 | ||
NM_015559.2 | NP_056374.2 | |||
AB007897.2 | ||||
AB022660.1 | BAA82444.1 | |||
BC062338.1 | AAH62338.1 | |||
BC146776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2272 | Chr.18:44678516 - 44723338 on Build GRCh38 | Deletion | SETBP1 LOC101927943 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |