Assay Details
Target Gene Details
Entrez Gene ID: | 56984 |
Gene Name: | proteasome assembly chaperone 2 |
Gene Aliases: |
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1 |
Location: |
Chr.18:12658739-12725740 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG2 | NM_147163.1 | NP_671692.1 | ||
AF068295.1 | AAF65184.1 | |||
AF161378.1 | AAF28938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576507 | Chr.18:12657196 - 12673188 on Build GRCh38 | Loss | SPIRE1 CEP76 PSMG2 |
nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss | SPIRE1 TUBB6 LOC100996333 PRELID3A ANKRD62 AFG3L2 CIDEA C18orf61 LOC105371998 CEP76 LOC100996324 PTPN2 PSMG2 |
esv2716814 | Chr.18:12641445 - 12668049 on Build GRCh38 | Deletion | SPIRE1 CEP76 PSMG2 |
nsv523743 | Chr.18:12474506 - 12742810 on Build GRCh38 | Loss | SPIRE1 CEP76 LOC100996324 PSMG2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |