Assay Details
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)