Assay Details
Target Gene Details
Entrez Gene ID: | 9480 |
Gene Name: | one cut homeobox 2 |
Gene Aliases: |
OC-2, OC2 |
Location: |
Chr.18:57435420-57491298 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ONECUT2 | NM_004852.2 | NP_004843.2 | ||
XM_017026096.1 | XP_016881585.1 | |||
Y18198.1 | CAB38253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828250 | Chr.18:57424010 - 57442352 on Build GRCh38 | Gain | ONECUT2 |
nsv953552 | Chr.18:57429769 - 57447868 on Build GRCh38 | Deletion | ONECUT2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |