Assay Details
Target Gene Details
Entrez Gene ID: | 7525 |
Gene Name: | YES proto-oncogene 1, Src family tyrosine kinase |
Gene Aliases: |
HsT441, P61-YES, Yes, c-yes |
Location: |
Chr.18:721592-812326 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YES1 | NM_005433.3 | NP_005424.1 | ||
XM_005258139.3 | XP_005258196.1 | |||
XM_017025960.1 | XP_016881449.1 | |||
AK314063.1 | ||||
BC048960.1 | AAH48960.1 | |||
M15990.1 | AAA35735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576236 | Chr.18:319176 - 794272 on Build GRCh38 | Gain | CETN1 ENOSF1 TYMSOS COLEC12 TYMS YES1 CLUL1 LOC105376854 |
dgv5810n54 | Chr.18:351004 - 2238246 on Build GRCh38 | Gain | CETN1 LOC388456 LINC00470 ENOSF1 TYMSOS COLEC12 ADCYAP1 TYMS YES1 CLUL1 LOC105376854 |
nsv576242 | Chr.18:567127 - 841320 on Build GRCh38 | Gain | CETN1 ENOSF1 TYMSOS TYMS YES1 CLUL1 |
nsv528169 | Chr.18:551688 - 2240220 on Build GRCh38 | Loss | CETN1 LOC388456 LINC00470 ENOSF1 TYMSOS ADCYAP1 TYMS YES1 CLUL1 |
nsv1060188 | Chr.18:575087 - 863351 on Build GRCh38 | Gain | CETN1 ENOSF1 TYMSOS TYMS YES1 CLUL1 |
nsv1057808 | Chr.18:766118 - 803577 on Build GRCh38 | Loss | YES1 |
esv2761984 | Chr.18:724222 - 855263 on Build GRCh38 | Loss | YES1 |
nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss | THOC1 LINC00470 TYMSOS NDC80 MIR8078 USP14 ROCK1P1 METTL4 CLUL1 CETN1 LOC388456 ENOSF1 COLEC12 ADCYAP1 EMILIN2 LOC101926997 CBX3P2 TYMS YES1 LOC105376854 SMCHD1 |
nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss | THOC1 LINC00470 TYMSOS MIR8078 USP14 ROCK1P1 CLUL1 CETN1 LOC388456 ENOSF1 COLEC12 ADCYAP1 TYMS YES1 LOC105376854 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |