Assay Details
Target Gene Details
Entrez Gene ID: | 9811 |
Gene Name: | cap binding complex dependent translation initiation factor |
Gene Aliases: |
Gm672, KIAA0427 |
Location: |
Chr.18:48539056-48863217 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTIF | NM_001142397.1 | NP_001135869.1 | ||
NM_014772.2 | NP_055587.1 | |||
XM_005258392.4 | XP_005258449.1 | |||
XM_006722583.3 | XP_006722646.1 | |||
XM_006722586.3 | XP_006722649.1 | |||
XM_006722587.3 | XP_006722650.1 | |||
XM_006722588.3 | XP_006722651.1 | |||
XM_006722589.3 | XP_006722652.1 | |||
XM_011526279.2 | XP_011524581.1 | |||
XM_017026100.1 | XP_016881589.1 | |||
XM_017026101.1 | XP_016881590.1 | |||
XM_017026102.1 | XP_016881591.1 | |||
AB007887.1 | ||||
BC042146.1 | AAH42146.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576855 | Chr.18:48549725 - 48615968 on Build GRCh38 | Loss |
![]() |
nsv833642 | Chr.18:48449936 - 48694680 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)