Assay Details
Target Gene Details
Entrez Gene ID: | 55520 |
Gene Name: | elaC ribonuclease Z 1 |
Gene Aliases: |
D29 |
Location: |
Chr.18:50968017-50988120 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELAC1 | NM_018696.2 | NP_061166.1 | ||
AB029151.1 | BAA96799.1 | |||
AF308695.1 | AAG24917.1 | |||
AI870985.1 | ||||
AK024822.1 | BAB15021.1 | |||
AK223507.1 | BAD97227.1 | |||
AK310911.1 | ||||
BC014624.2 | AAH14624.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |