Assay Details
Target Gene Details
Entrez Gene ID: | 1005 |
Gene Name: | cadherin 7 |
Gene Aliases: |
CDH7L1 |
Location: |
Chr.18:65750252-65885136 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH7 | NM_001317214.1 | NP_001304143.1 | ||
NM_004361.3 | NP_004352.2 | |||
NM_033646.2 | NP_387450.1 | |||
XM_017025523.1 | XP_016881012.1 | |||
AB035301.1 | BAA87415.1 | |||
AJ007611.1 | CAC13127.1 | |||
BC036786.1 | AAH36786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1067311 | Chr.18:63818011 - 66637685 on Build GRCh38 | Loss | LINC01538 CDH7 HMSD CDH19 SERPINB2 LOC284294 LINC00305 SERPINB10 SERPINB8 |
nsv833675 | Chr.18:65631834 - 65807471 on Build GRCh38 | Gain | CDH7 |
nsv458102 | Chr.18:65715200 - 68002467 on Build GRCh38 | Loss | CDH7 CDH19 DSEL LOC643542 LOC101927430 MIR5011 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |