Assay Details
Target Gene Details
Entrez Gene ID: | 147323 |
Gene Name: | StAR related lipid transfer domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.18:54324592-54357890 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD6 | NM_139171.1 | NP_631910.1 | ||
XM_011525821.1 | XP_011524123.1 | |||
XM_017025552.1 | XP_016881041.1 | |||
XM_017025553.1 | XP_016881042.1 | |||
XM_017025554.1 | XP_016881043.1 | |||
AF480305.1 | AAL89655.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)