Assay Details
Target Gene Details
Entrez Gene ID: | 101927698 |
Gene Name: | DSC1/DSC2 antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.18:31101588-31162789 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DSCAS | NR_110785.1 | |||
DA156712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519734 | Chr.18:31127452 - 31141571 on Build GRCh38 | Loss | DSCAS DSC1 |