Assay Details
Target Gene Details
Entrez Gene ID: | 317719 |
Gene Name: | kelch like family member 10 |
Gene Aliases: |
SPGF11 |
Location: |
Chr.17:41835685-41848347 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLHL10 | NM_152467.3 | NP_689680.2 | ||
XM_011524706.2 | XP_011523008.1 | |||
AK057224.1 | BAB71387.1 | |||
AK301797.1 | ||||
AK302141.1 | ||||
AY495339.1 | AAS91792.1 | |||
BP371285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3174n100 | Chr.17:41778258 - 41843183 on Build GRCh38 | Gain | JUP NT5C3B FKBP10 KLHL10 P3H4 |
nsv1057008 | Chr.17:41832887 - 41885423 on Build GRCh38 | Gain | NT5C3B KLHL10 KLHL11 ACLY |
nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss | NT5C3B FKBP10 KLHL10 KLHL11 P3H4 ACLY TTC25 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |