Assay Details
Target Gene Details
Entrez Gene ID: | 51268 |
Gene Name: | pipecolic acid and sarcosine oxidase |
Gene Aliases: |
LPIPOX |
Location: |
Chr.17:29042900-29057227 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIPOX | NM_016518.2 | NP_057602.2 | ||
XM_017024722.1 | XP_016880211.1 | |||
AF134593.1 | AAF37331.1 | |||
AF136970.1 | AAG49431.1 | |||
AK022773.1 | ||||
AK027498.1 | ||||
BC008960.2 | AAH08960.1 | |||
BC027622.2 | AAH27622.1 | |||
CR457155.1 | CAG33436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833408 | Chr.17:28913470 - 29105450 on Build GRCh38 | Loss | TIAF1 MYO18A LOC101927018 PIPOX SEZ6 PHF12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |