Assay Details
Target Gene Details
Entrez Gene ID: | 80279 |
Gene Name: | CDK5 regulatory subunit associated protein 3 |
Gene Aliases: |
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553 |
Location: |
Chr.17:47967904-47981786 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDK5RAP3 | NM_001278197.1 | NP_001265126.1 | ||
NM_001278198.1 | NP_001265127.1 | |||
NM_001278216.1 | NP_001265145.1 | |||
NM_001278217.1 | NP_001265146.1 | |||
NM_176096.2 | NP_788276.1 | |||
XM_011525297.1 | XP_011523599.1 | |||
XM_017025164.1 | XP_016880653.1 | |||
XM_017025165.1 | XP_016880654.1 | |||
XM_017025166.1 | XP_016880655.1 | |||
AB062433.1 | BAB93496.1 | |||
AF110322.1 | AAG39277.1 | |||
AF217982.2 | AAG17225.1 | |||
AF343090.1 | AAK69655.1 | |||
AK023722.1 | BAB14657.1 | |||
AK223387.1 | BAD97107.1 | |||
AK225274.1 | ||||
AK300643.1 | ||||
BC009957.2 | AAH09957.1 | |||
BC072435.1 | AAH72435.1 | |||
DA410409.1 | ||||
DA410477.1 | ||||
DB075547.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |