Assay Details
Target Gene Details
Entrez Gene ID: | 3690 |
Gene Name: | integrin subunit beta 3 |
Gene Aliases: |
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT |
Location: |
Chr.17:47253842-47312711 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB3 | NM_000212.2 | NP_000203.2 | ||
BC127666.1 | ||||
BC127667.1 | ||||
J02703.1 | AAA52589.1 | |||
KC120777.1 | ||||
KC120778.1 | ||||
M20311.1 | AAA60122.1 | |||
M25108.1 | AAA36121.1 | |||
M35999.1 | AAA35927.1 | |||
U95204.1 | AAB71380.1 |
Target Gene Details
Entrez Gene ID: | 102724508 |
Gene Name: | thyroid cancer-associated transcript 158 |
Gene Aliases: |
- |
Location: |
Chr.17:47303443-47323901 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THCAT158 | NR_110880.1 | |||
NR_110881.1 | ||||
AA430297.1 | ||||
AA609236.1 | ||||
AW293117.1 | ||||
HY036509.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |