Assay Details
Target Gene Details
Entrez Gene ID: | 8456 |
Gene Name: | forkhead box N1 |
Gene Aliases: |
FKHL20, RONU, WHN |
Location: |
Chr.17:28506211-28538900 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXN1 | XM_005258046.3 | XP_005258103.1 | ||
XM_011525358.1 | XP_011523660.1 | |||
XM_011525359.1 | XP_011523661.1 | |||
XM_011525362.1 | XP_011523664.1 | |||
XM_011525368.2 | XP_011523670.1 | |||
XM_017025228.1 | XP_016880717.1 | |||
XM_017025229.1 | XP_016880718.1 | |||
XM_017025230.1 | XP_016880719.1 | |||
XM_017025231.1 | XP_016880720.1 | |||
AK313878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064958 | Chr.17:28490519 - 28565886 on Build GRCh38 | Loss | UNC119 SLC13A2 PIGS FOXN1 |
nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain | UNC119 ALDOC SNORD4B TLCD1 FOXN1 PROCA1 SGK494 SUPT6H SPAG5-AS1 RAB34 RPL23A RPS7P1 SPAG5 KIAA0100 SNORD4A NEK8 SLC13A2 TRAF4 PIGS NARR SDF2 SNORD42B SNORD42A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |