Assay Details
Target Gene Details
Entrez Gene ID: | 10160 |
Gene Name: | FERM, ARH/RhoGEF and pleckstrin domain protein 1 |
Gene Aliases: |
CDEP, FARP1-IT1, PLEKHC2, PPP1R75 |
Location: |
Chr.13:98142562-98449773 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FARP1 | NM_001001715.3 | NP_001001715.2 | ||
NM_001286839.1 | NP_001273768.1 | |||
NM_005766.3 | NP_005757.1 | |||
XM_011521046.2 | XP_011519348.1 | |||
XM_017020312.1 | XP_016875801.1 | |||
AB008430.1 | BAA24267.1 | |||
AI273567.1 | ||||
AI274149.1 | ||||
BC041595.1 | AAH41595.1 | |||
BC071592.1 | AAH71592.1 | |||
BM763139.1 | ||||
CF994038.1 | ||||
DB498534.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |