Assay Details
Target Gene Details
Entrez Gene ID: | 84945 |
Gene Name: | abhydrolase domain containing 13 |
Gene Aliases: |
BEM46L1, C13orf6, bA153I24.2 |
Location: |
Chr.13:108218415-108235440 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD13 | NM_032859.2 | NP_116248.2 | ||
XM_011521128.2 | XP_011519430.1 | |||
BC022566.1 | AAH22566.2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |