Assay Details
Target Gene Details
Entrez Gene ID: | 100861550 |
Gene Name: | PDX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:27829759-27921404 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDX1-AS1 | NR_047484.1 | 4 | 1809 | |
AW340746.1 | 1 | 196 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv977221 | Chr.13:27828708 - 27829994 on Build GRCh38 | Duplication | PDX1-AS1 |